Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A6977

Sigma-Aldrich

Anti-ADAM-9, N-Terminal Active antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-A Disintegrin And Metalloproteinase-9, Anti-MDC9, Anti-Meltrin-γ

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

human

concentration

~1 mg/mL

technique(s)

western blot: 1:1,000

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ADAM9(8754)

General description

ADAM9 (MDC9 or Meltrin-gamma) belongs to the metalloproteinase family that has disintegrin-like domains (ADAMs). ADAM9 is widely expressed in tissues such as heart, lung and brain cells. This metalloproteinase regulates myoblast functions and activates TNF-α (tumor necrosis factor-α) and HB-EGF. ADAM9 also functions as an adhesion protein with integrins αvβ5 and α6β1.
Rabbit Anti-ADAM-9, N-Terminal Active antibody binds to the N-terminus of active human ADAM9.
By immunoblotting against the reduced protein, the antibody identifies bands at 116 kDa (minor band), 66 kDa (major band), and breakdown products at 55 kDa and 34 kDa in cell lysates.

Immunogen

synthetic peptide corresponding to the N-terminus of active human ADAM9.

Application

Rabbit Anti-ADAM-9, N-Terminal Active antibody can be used for western blot applications at a dilution of 1:1,000.

Physical form

Solution in phosphate buffered saline containing 50% glycerol and 0.05% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Zhou et al.
Biochemical and biophysical research communications, 280(2), 574-580 (2001-02-13)
MDC-9 is a widely expressed member of the metalloproteinase/disintegrin/cysteine-rich protein family. The disintegrin domain of MDC-9 lacks an RGD motif but has recently been reported to bind the alpha(6)beta(1) integrin; however, it is unclear whether MDC-9 can bind other integrins.
D Nath et al.
Journal of cell science, 113 ( Pt 12), 2319-2328 (2000-05-29)
The ADAMs (A Disintegrin and Metalloprotease Domains) are a family of membrane-anchored proteins that play a role in fertilisation, myoblast fusion and ectodomain shedding of cell surface proteins. Meltrin gamma (ADAM-9) is a widely expressed member of this family and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service