Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A6728

Sigma-Aldrich

β1-Adrenergic Receptor preparation human

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202

biological source

human

form

liquid

technique(s)

drug transporter assay: suitable
enzyme immunoassay: suitable

UniProt accession no.

storage temp.

−70°C

Gene Information

human ... ADRB1(153)

Application

The β1-adrenergic receptor is the classical target for β-blockers class of cardiovascular drugs. It is also involved in regulation of lipolysis in adipocytes, where its overexpression is seen to confer partial resistance to diet-induced obesity.

Biochem/physiol Actions

Membranes suspended in 50 mM Tris-HCl containing 12 mM MgCl2 and 2 mM EDTA, pH 7.4. Actual concentration and specific binding capacity are provided with each lot.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service