A6605
AMN082
≥98% (HPLC), solid
Synonym(s):
N,N′-Dibenzhydrylethane-1,2-diamine dihydrochloride
About This Item
assay
≥98% (HPLC)
form
solid
storage condition
desiccated
color
white to off-white
solubility
DMSO: ≥5 mg/mL
storage temp.
−20°C
SMILES string
Cl[H].Cl[H].C(CNC(c1ccccc1)c2ccccc2)NC(c3ccccc3)c4ccccc4
InChI
1S/C28H28N2.2ClH/c1-5-13-23(14-6-1)27(24-15-7-2-8-16-24)29-21-22-30-28(25-17-9-3-10-18-25)26-19-11-4-12-20-26;;/h1-20,27-30H,21-22H2;2*1H
InChI key
YRQCDCNQANSUPB-UHFFFAOYSA-N
Application
Biochem/physiol Actions
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Faceshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Sigma-Aldrich offers many products related to G-protein family glutamate receptors for your research needs.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service