Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A6487

Sigma-Aldrich

Alpha-Lytic Protease M190A Mutant

Synonym(s):

Alpha-Lytic Protease M190A Mutant, ALP M190A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
NACRES:
NA.54

recombinant

expressed in E. coli

form

liquid

specific activity

≥0.05 U/mg

mol wt

19.8 kDa (mature form)

optimum pH

5.0(storage)
7.5(activity)

pI 

9.69

shipped in

dry ice

Storage temp.

−70°C

General description

Alpha-lytic protease (aLP) is an alternative specificity protease for proteomics applications, whose wild-type (WT) version cleaves after T, A, S, and V residues. The M190A (Met190 → Ala190) mutant of aLP has different cleavage specificities, and cleaves after M, F, and L residues. Both the WT and M190A forms of aLP generate peptides of similar average length as trypsin.

In WT aLP, the methionines at positions 190 and 213 cause WT aTP to have its particular specificity toward peptide substrates with small hydrophobic side chains at the P1 position. The M190A mutation gives the resulting mutant M190A aLP the ability to cleave peptide substrates with large hydrophobic side chains at the P1 side chain with greater efficiency compared to WT aLP, in addition to accommodating peptides with small hydrophobic side chains as before. Other structural changes in M190A aLP compared to WT aLP have been discussed.

The activity of M190A aLP in the presence of various solution components is as follows:
  • 0.1% sodium deoxycholate: ~1.4-fold enhanced activity
  • 1.0% sodium deoxycholate: nearly full activity
  • 0.1% SDS: ~40% activity
  • 1.0% SDS: ~30% activity
  • 1 M guanidine HCl: ~20% activity
  • 4 M guanidine HCl: ~1% activity (essentially inactivated)

Unit Definition

mmols of pNA produced per min per mg protein from 0.5 mM N-succinyl-Ala-Ala-Pro-Phe-PNA at 25 °C in 100 mM Tris-HCl (pH 7.5).

related product

Product No.
Description
Pricing

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J E Mace et al.
Journal of molecular biology, 254(4), 720-736 (1995-12-08)
Gly216 in the active site of the broadly specific MA190 mutant of alpha-lytic protease has been found to be remarkably tolerant of amino acid substitutions. Side-chains as large as Trp can be accommodated within the substrate-binding pocket without abolishing catalysis
D W Miller et al.
Journal of molecular biology, 286(1), 267-278 (1999-02-05)
We have used alpha-lytic protease as a model system for exploring the relationship between the internal dynamics of an enzyme and its substrate specificity. The wild-type enzyme is highly specific for small substrates in its primary specificity pocket, while the
R Bone et al.
Nature, 339(6221), 191-195 (1989-05-18)
The substrate specificity of alpha-lytic protease has been changed dramatically, with a concomitant increase in activity, by replacing an active-site Met with Ala. The substrate specificity of both this mutant and another similar mutant are extraordinarily broad. X-ray crystallographic analysis
The α-Lytic Protease.
Whitaker, D.R.
Methods in Enzymology, 19, 599-613 (1970)
Jesse G Meyer et al.
Molecular & cellular proteomics : MCP, 13(3), 823-835 (2014-01-16)
Bottom-up proteomics studies traditionally involve proteome digestion with a single protease, trypsin. However, trypsin alone does not generate peptides that encompass the entire proteome. Alternative proteases have been explored, but most have specificity for charged amino acid side chains. Therefore

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service