A6481
Allene Oxide Synthase from Parthenium argentatum
recombinant, expressed in E. coli, 25,000-40,000 units/mg protein
Synonym(s):
AOS, Allene Oxide Synthase Parthenium argentatum
About This Item
recombinant
expressed in E. coli
form
solution
specific activity
25,000-40,000 units/mg protein
packaging
vial of ~300 μg
shipped in
dry ice
storage temp.
−20°C
Application
Biochem/physiol Actions
Unit Definition
Physical form
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service