Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A6352

Sigma-Aldrich

Anti-ADAMTS-2, Propeptide Region antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-A Disintegrin And Metalloproteinase with Thrombospondin-2 motif, Anti-Procollagen I N-Proteinase

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

human

concentration

~1 mg/mL

technique(s)

western blot: 1:1,000

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ADAMTS2(9509)

General description

ADAMTS-2 (or Procollagen I N-Proteinase, PNP) belongs to the ADAM (A Disintegrin And Metalloproteinase) family and has thrombospondin (TS) repeats. ADAMTS-2 has been implicated in collagen synthesis, angiogenesis and development. Alterations in this gene have been linked to Ehlers-Danlos syndrome type VII C, dermatosparaxis, and male infertility. Rabbit Anti-ADAMTS-2 binds to human ADAMTS-2.
By immunoblotting against the reduced protein, the antibody identifies the zymogen form at 130-140 kDa, activated forms at 94-105 kDa (major bands), and breakdown products at 50 kDa, 34 kDa in cell culture media and lysates.

Immunogen

synthetic peptide corresponding to the propeptide domain of the human ADAMTS-2.

Application

Rabbit Anti-ADAMTS-2 can be used for western blot applications at a dilution of 1:1,000.

Physical form

Solution in phosphate buffered saline containing 50% glycerol and 0.05% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Colige et al.
Proceedings of the National Academy of Sciences of the United States of America, 94(6), 2374-2379 (1997-03-18)
Procollagen N-proteinase (EC 3.4.24.14) cleaves the amino-propeptides in the processing of type I and type II procollagens to collagens. Deficiencies of the enzyme cause dermatosparaxis in cattle and sheep, and they cause type VIIC Ehlers-Danlos syndrome in humans, heritable disorders
B L Tang
The international journal of biochemistry & cell biology, 33(1), 33-44 (2001-02-13)
ADAMTS (a disintegrin and metalloprotease with thrombospondin motifs) is a novel family of extracellular proteases found in both mammals and invertebrates. Members of the family may be distinguished from the ADAM (a disintegrin and metalloprotease) family members based on the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service