A5973
Amyloid-β-Protein Fragment 1-40, All D-Amino Acids
>70% (HPLC), lyophilized powder
Synonym(s):
β-Amyloid (Aβ)[1-40, all D-amino acids]
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
MDL number:
UNSPSC Code:
12352200
assay
>70% (HPLC)
form
lyophilized powder
mol wt
4331
composition
Protein content, ~80%
solubility
H2O: soluble
storage temp.
−20°C
Gene Information
human ... APP(351)
mouse ... APP(11820)
rat ... APP(54226)
Amino Acid Sequence
Asp-Ala-Glu-Phe-Arg-His-Asp-Ser-Gly-Tyr-Glu-Val-His-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Glu-Asp-Val-Gly-Ser-Asn-Lys-Gly-Ala-Ile-Ile-Gly-Leu-Met-Val-Gly-Gly-Val-Val
Application
Used in the study of plaques and tangles that occur in Alzheimer′s disease (AD) patients.
Biochem/physiol Actions
This D-amino acid peptide functions as a control useful in elucidating structural dependence of aggregation properties characteristic of the amyloid-β 1-40 peptide associated with plaque formation and Alzheimer′s disease.
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service