Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A5851

Sigma-Aldrich

Anti-ASK-1, C-terminal antibody produced in rabbit

~0.5 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Apoptosis Signal-Regulating Kinase 1, Anti-MAPKKK5

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.44

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 155 kDa

species reactivity

human

concentration

~0.5 mg/mL

technique(s)

western blot: 0.5-1.0 μg/mL using SW1353 human chondrosarcoma whole cell lysate.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... MAP3K5(4217)

General description

ASK1 (Apoptosis Signal-regulating Kinase 1) also known as MAPKKK5 (Mitogen activated protein kinase kinase kinase 5) activates JNK and p38 signaling cascades. ASK-1 is activated in response to stress such as UV and X-ray radiation, proinflammatory cytokines such as TNF-α, heat shock or osmotic shock. Overexpression of ASK1 induces apoptotic cell death. Other signalling molecules such as PP5 and reactive oxygen species regulate the pro-apoptotic signalling by ASK1 hence act as negative regulators. ASK1 has pivotal function also in a broad range of biological activities and aids the cells in adapting to varied stress factors and in induction of immune responses.
Anti-ASK1 specifically recognizes human ASK1 (155 kDa).

Immunogen

synthetic peptide corresponding to amino acids 1356-1375 of the human C-terminal region of ASK1.

Application

A concentration of 1 μg/mL of anti-ASK-1 is recommended for detection by immunoblotting in whole cell extract of SW1353 human chondrosarcoma cells.

Quantity

Reagent is sufficient for 10 blots.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kohsuke Takeda et al.
Cell structure and function, 28(1), 23-29 (2003-03-26)
Apoptosis signal-regulating kinase 1 (ASK1) is a ubiquitously expressed mitogen-activated protein (MAP) kinase kinase kinase that activates the c-Jun N-terminal kinase (JNK) and p38 MAP kinase signaling cascades. Recent findings from analyses of ASK1-deficient mice have revealed that ASK1 is
Jun Matsukawa et al.
Journal of biochemistry, 136(3), 261-265 (2004-12-16)
The mitogen-activated protein (MAP) kinase cascades play essential roles in a variety of cell processes by influencing transcriptional or translational regulation. ERKs play a central role in survival and mitogenic signaling, while JNKs and p38 MAP kinases are preferentially activated
H Ichijo et al.
Science (New York, N.Y.), 275(5296), 90-94 (1997-01-03)
Mitogen-activated protein (MAP) kinase cascades are activated in response to various extracellular stimuli, including growth factors and environmental stresses. A MAP kinase kinase kinase (MAPKKK), termed ASK1, was identified that activated two different subgroups of MAP kinase kinases (MAPKK), SEK1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service