Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A5104

Sigma-Aldrich

Anti-Water Channel Aquaporin 6 antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

mouse, rat

technique(s)

western blot: 1:200 using rat kidney membranes

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

mouse ... Aqp6(11831)
rat ... Aqp6(29170)

General description

AQP6 is part of a family of Aquaporins that function to transport water across cell membranes. AQP6 is believed to be kidney-specific.
Rabbit Anti-Water Channel Aquaporin 6 (AQP6) antibody binds mouse and rat AQP6.

Immunogen

peptide corresponding to residues 259-276 of rat AQP6. Mouse is 15/18 residues identical.

Application

Rabbit Anti-Water Channel Aquaporin 6 antibody can be used for western blot applications at a dilution of 1:200 using rat kidney membrane extracts.

Physical form

Lyophilized from phosphate buffered saline, pH 7.4, 0.025% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service