A5101
Anti-ADAM-9, Propeptide Domain antibody produced in rabbit
~1 mg/mL, affinity isolated antibody, buffered aqueous glycerol solution
Synonym(s):
Anti-A Disintegrin And Metalloproteinase-9, Anti-MDC9, Anti-Meltrin-γ
About This Item
biological source
rabbit
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
form
buffered aqueous glycerol solution
species reactivity
human, rat, pig
concentration
~1 mg/mL
technique(s)
western blot: 1:1,000
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... ADAM9(8754)
rat ... Adam9(290834)
General description
Anti-ADAM-9, Propeptide Domain may be used for the detection and localization of human ADAM9 and does not react with other ADAMs. The antibody is also reacts with rat, pig and human ADAM9.
Immunogen
Application
Physical form
Not finding the right product?
Try our Product Selector Tool.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service