Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A4986

Sigma-Aldrich

PASK (981-end), active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

DKFZp434O051, DKFZp686P2031, KIAA0135, PASKIN, STK37

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO® Kinase

assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

122-166 nmol/min·mg

mol wt

~66 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PASK(23178)

Biochem/physiol Actions

PASK or PAS domain containing serine/threonine kinase regulates the function of many intracellular signaling pathways involved in stress. PASK is involved in sensing environmental changes in light intensity, oxygen concentration and redox potential. Through interaction with IRS-1, PASK has been proposed as a counter-regulatory mechanism in insulin and cytokine signaling. PASK can phosphorylate and inactivate glycogen synthase in vitro. Efficient phosphorylation requires residues 444 to 955 of PASK between the PAS and catalytic kinase domain and this interaction is inhibited by the PAS domain.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.25 mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Nagase et al.
DNA research : an international journal for rapid publication of reports on genes and genomes, 2(4), 167-174 (1995-08-31)
In this series of projects regarding the accumulation of sequence information of unidentified human genes, we newly deduced the sequences of 40 full-length cDNA clones of human cell line KG-1, and predicted the coding sequences of the corresponding genes, named

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service