Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A4793

Sigma-Aldrich

Adenosine 5′-triphosphate–Agarose

aqueous glycerol suspension

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41106500
NACRES:
NA.56

form

aqueous glycerol suspension

extent of labeling

1-3 μmol per mL

matrix

cross-linked 4% beaded agarose

matrix activation

epoxy

matrix attachment

ribose hydroxyls

matrix spacer

22 atoms (adipic acid dihydrazide)

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Adenosine 5′-triphosphate Agarose (5′-ATP agarose) has been used in affinity chromatography to purify uridine kinase from Ehrlich ascites tumor cells.

Physical form

Suspension in 50% glycerol containing preservative

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Patrick C Fraering et al.
The Journal of biological chemistry, 280(51), 41987-41996 (2005-10-21)
gamma-Secretase is an unusual protease with an intramembrane catalytic site that cleaves many type I membrane proteins, including the amyloid beta-protein (Abeta) precursor (APP) and the Notch receptor. Genetic and biochemical studies have identified four membrane proteins as components of
K Nadeau et al.
Protein science : a publication of the Protein Society, 1(8), 970-979 (1992-08-01)
A Crithidia fasciculata 83-kDa protein purified during a separate study of C. fasciculata trypanothione synthetase was shown to have ATPase activity and to belong to the hsp90 family of stress proteins. Because no ATPase activity has previously been reported for
P Csermely et al.
The Journal of biological chemistry, 266(8), 4943-4950 (1991-03-15)
The 90-kDa heat shock protein (hsp-90) is an abundant cytosolic protein believed to play a role in maintenance of protein trafficking and closely associated with several steroid hormone receptors. Incubation of highly purified hsp-90 with [gamma-32P]ATP results in its autophosphorylation
D R Drummond et al.
European journal of biochemistry, 209(1), 171-179 (1992-10-01)
Twenty-five mutations were created in the Drosophila melanogaster Act88F actin gene by in vitro mutagenesis and the mutant actins expressed in vitro. The affinity of the mutant actins for ATP, profilin and DNase I was determined. They were also tested
Purification and characterization of acetate kinase from <I>Clostridium thermocellum</I>.
Lin, W.R., et al.
Tetrahedron, 54(52), 15915-15925 (1998)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service