Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A4729

Sigma-Aldrich

Anti-β2 Adrenergic Receptor antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

availability

not available in Japan

technique(s)

immunocytochemistry: 5 μg/mL using human heart tissue
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 5 μg/mL using human heart tissue

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ADRB2(154)
mouse ... Adrb2(11555)
rat ... Adrb2(24176)

General description

β2 Adrenergic Receptor encodes beta-2-adrenergic receptor which is a member of the G protein-coupled receptor superfamily. This receptor is directly associated with one of its ultimate effectors, the class C L-type calcium channel Ca(V)1.2. This rec
Rabbit Anti-β2 Adrenergic Receptor antibody binds to β2 adrenergic receptor in human heart tissues.

Immunogen

synthetic peptide corresponding to the C-terminal domain of human β2 adrenergic receptor. The immunizing peptide has 83% homology with the rat gene and 67% homology with the mouse gene.

Application

Rabbit Anti-β2 Adrenergic Receptor antibody can be used for immunocytochemistry and immunohistochemistry applications at 5μg/ml using human heart tissues.

Target description

Adrenergic receptors are a G-protein coupled receptors that exist in two forms, α or β. Ligands for adrenergic receptors are catecholamines that induce a sympathetic response in animal cells.

Physical form

Solution in phosphate buffered saline, containing 0.1% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service