Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A4720

Sigma-Aldrich

Monoclonal Anti-Phosphotyrosine−Agarose antibody produced in mouse

clone PY20, purified immunoglobulin, buffered aqueous suspension

Synonym(s):

Monoclonal Anti-Phosphotyrosine, Phospho-Tyr, Phospho-tyrosine, p-Tyr

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

mouse

conjugate

agarose conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

PY20, monoclonal

form

buffered aqueous suspension

technique(s)

immunoprecipitation (IP): suitable

isotype

IgG2b

shipped in

wet ice

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Reversible phosphorylation of proteins is an important post-translational modification that plays a regulatory role in the expression of most proteins in the cells. Reversible phosphorylation at multiple serine, tyrosine and threonine residues mediates numerous signalling pathways in both prokaryotic and eukaryotic cells . Cellular proteins with phosphorylated tyrosine increase many fold by the activation of tyrosine kinases. Most mitogenic receptor systems such as EGF, PDGF, insulin receptors contain serine/threonine/tyrosine kinase domains that undergo autophosphorylation when receptors bind to the respective ligands. Monoclonal anti-phosphotyrosine−agarose antibody can be used in immunoprecipitation. Mouse anti-phosphotyrosine−agarose antibody reacts specifically with both native and denatured proteins comprising phosphorylated tyrosine.

Immunogen

phosphotyrosine-protein conjugate

Application

Monoclonal anti-phosphotyrosine−agarose antibody can be used in immunoaffinity purification of phosphotyrosyl proteins, including transforming proteins, steroid hormone receptors (e.g., estradiol and glucocorticoid receptors), growth factor receptors (e.g., EGF and PDGF receptors), insulin receptors and other proteins involved in the control of cellular growth.

Physical form

Suspension in 20 mM sodium phosphate, pH 7.5, 150 mM sodium chloride, 3 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vanessa Liévin-Le Moal et al.
Cellular microbiology, 13(5), 764-785 (2011-03-01)
CEACAM1 expressed by granulocytes and epithelial cells is recognized as a membrane-associated receptor by some Gram-negative pathogens. Here we report a previously unsuspected role of human CEACAM1-4L (hCEACAM1-4L) in polarized epithelial cells. We find that in contrast with non-transfected cells
A Dvir et al.
The Journal of cell biology, 113(4), 857-865 (1991-05-01)
Protein tyrosine kinase blockers of the tyrphostin family inhibited the EGF-dependent proliferation of human and guinea pig keratinocytes grown in culture and induced their growth arrest. These blockers also significantly inhibited the growth of epidermal keratinocytes, but not of dermal
Wiljan J A J Hendriks et al.
The FEBS journal, 275(5), 816-830 (2008-02-27)
Some 40-odd genes in mammals encode phosphotyrosine-specific, 'classical' protein tyrosine phosphatases. The generation of animal model systems and the study of various human disease states have begun to elucidate the important and diverse roles of protein tyrosine phosphatases in cellular
Joanna Cieśla et al.
Acta biochimica Polonica, 58(2), 137-148 (2011-05-31)
Reversible phosphorylation is the most widespread posttranslational protein modification, playing regulatory role in almost every aspect of cell life. The majority of protein phosphorylation research has been focused on serine, threonine and tyrosine that form acid-stable phosphomonoesters. However, protein histidine

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service