Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A4351

Sigma-Aldrich

Anti-ADAM-17, Cytoplasmic Domain antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-A Disintegrin And Metalloproteinase-17, Anti-TACE, Anti-TNF-α Converting Enzyme

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

mol wt

antigen 110 kDa

species reactivity

pig, rat, human

concentration

~1 mg/mL

technique(s)

western blot: 1:1,000

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ADAM17(6868)
rat ... Adam17(57027)

General description

ADAM-17 or tumor necrosis factor converting enzyme (TACE) is known to regulate several cellular functions such as cell growth and movement. It has a varied array of substrates that include adhesion molecules, growth factors and their receptors, as well as many cytokines. Thus, ADAM-17 abnormalities have been associated with various disorders such as renal fibrosis, Alzheimer′s disease, rheumatoid arthritis, cancer, diabetes and cardiac disorders.
Rabbit anti-ADAM-17, cytoplasmic domain antibody recognizes human, porcine, and rat ADAM17. It does not react with other ADAMs. By immunoblotting against the reduced protein, the antibody recognizes a band of 110kDa from conditioned media or cell lysates.

Immunogen

synthetic peptide corresponding to the cytoplasmic domain of full-length human ADAM-17.

Application

Rabbit anti-ADAM-17, cytoplasmic domain antibody can be used for western blot at a dilution of 1:1,000.

Physical form

Solution in 0.01 M phosphate buffered saline containing 50% glycerol and 0.05% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Monika Gooz
Critical reviews in biochemistry and molecular biology, 45(2), 146-169 (2010-02-27)
This review focuses on the role of ADAM-17 in disease. Since its debut as the tumor necrosis factor converting enzyme (TACE), ADAM-17 has been reported to be an indispensible regulator of almost every cellular event from proliferation to migration. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service