Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A4348

Sigma-Aldrich

[Nle35]-Amyloid β-Protein Fragment 1-40

≥90% (HPLC), lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200

assay

≥90% (HPLC)

form

lyophilized powder

mol wt

4313

composition

Protein content, ≥75%

solubility

H2O: soluble

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... APP(351)

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L K Simmons et al.
Molecular pharmacology, 45(3), 373-379 (1994-03-01)
Amyloid beta peptide (A beta), the major protein constituent of senile plaques in patients with Alzheimer's disease, is believed to facilitate the progressive neurodegeneration that occurs in the latter stages of this disease. Early attempts to characterize the structure-activity relationship

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service