Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A4345

Sigma-Aldrich

Anti-E1A (Adenovirus Early Region 1A) antibody produced in sheep

fractionated antiserum

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

sheep

conjugate

unconjugated

antibody form

fractionated antiserum

antibody product type

primary antibodies

clone

polyclonal

form

PBS solution

mol wt

antigen 43 kDa

species reactivity

viral

technique(s)

western blot: 10 μg/mL using whole extract of 293 cells

shipped in

dry ice

Storage temp.

−20°C

target post-translational modification

unmodified

General description

Adenovirus E1A gene modulates gene expression by associating with transcription factors in cells. Studies have reported that E1A regulates the stimulation of ICAM-1 through inflammatory signals in pulmonary epithelial cells. E1A has also been implicated in chronic obstructive pulmonary disease (COPD). Furthermore, E1A is known to mediate tumor suppression by rendering epithelial cells sensitive to anoikis. Sheep Anti-E1A (Adenovirus Early Region 1A) antibody binds to E1A using immunoblotting (43kDa).

Immunogen

GST-E1A fusion protein.

Application

Anti-E1A (Adenovirus Early Region 1A) antibody can be used at 10μg/mL for western blot assays using whole extract of 293 cells.

Physical form

Solution in phosphate buffered saline containing 0.08% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S M Frisch
Mutation research, 350(1), 261-266 (1996-02-19)
Tumor suppressor genes such as Rb and p53 usually kill tumor cells when overexpressed ectopically. This is a consequence of their normal cell cycle regulatory functions. By contrast, the E1a gene of adenovirus, a common cold virus, converts tumor cells
N Keicho et al.
American journal of respiratory cell and molecular biology, 16(1), 23-30 (1997-01-01)
Previous studies from our laboratory demonstrated that adenovirus E1A DNA and proteins are detected in lungs of patients with chronic obstructive pulmonary disease (COPD). Since adenovirus E1A gene products are known to regulate the expression of many genes by interacting

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service