Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A4251

Sigma-Aldrich

N-Acetyl-L-phenylalanine ethyl ester

Synonym(s):

N-Ac-l-Phe-Oet, APAEE

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H17NO3
CAS Number:
Molecular Weight:
235.28
EC Number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.26

assay

≥98% (TLC)

form

powder

technique(s)

ligand binding assay: suitable

color

white

storage temp.

2-8°C

SMILES string

CCOC(=O)C(Cc1ccccc1)NC(C)=O

InChI

1S/C13H17NO3/c1-3-17-13(16)12(14-10(2)15)9-11-7-5-4-6-8-11/h4-8,12H,3,9H2,1-2H3,(H,14,15)

InChI key

YIVZYFDBEPMPNL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hua Zhao et al.
Journal of molecular catalysis. B, Enzymatic, 72(3-4), 163-167 (2011-09-13)
Deep eutectic solvents (DESs) consisting of mixtures of a choline salt (chloride or acetate form) and glycerol are prepared as easily accessible, biodegradable, and inexpensive alternatives to conventional aprotic cation-anion paired ionic liquids. These DES systems display excellent fluidity coupled
R L Levine et al.
Biochemistry, 21(11), 2600-2606 (1982-05-25)
The ultraviolet absorption spectrum of proteins in 6 M guanidine is approximately that of the sum of the spectra of the constituent aromatic amino acids, phenylalanine, tyrosine, and tryptophan, plus contributions from light scattering and disulfides. A multicomponent analysis of
Muhammad Nazir Tahir et al.
Bioprocess and biosystems engineering, 37(4), 687-695 (2013-08-28)
In this study, serine protease (subtilisin Carlsberg) was immobilized on pentynyl dextran (PyD, O-alkynyl ether of dextran, 1) and used for the transesterification of N-acetyl-L-phenylalanine ethyl ester (2) with different aliphatic (1-propanol, 1-butanol, 1-pentanol, 1-hexanol) and aromatic (benzyl alcohol, 2-phenyl
J A Laszlo et al.
Biotechnology and bioengineering, 75(2), 181-186 (2001-09-06)
The transesterification reaction of N-acetyl-L-phenylalanine ethyl ester with 1-propanol catalyzed by alpha-chymotrypsin was examined in the ionic liquids 1-butyl-3-methylimidazolium hexafluorophosphate ([bmim][PF(6)]) and 1-octyl-3-methylimidazolium hexafluorophosphate ([omim][PF(6)]), and in combination with supercritical carbon dioxide (SC-CO(2)). The activity of alpha-chymotrypsin was studied to
P C Quan et al.
Journal of immunology (Baltimore, Md. : 1950), 128(4), 1786-1791 (1982-04-01)
The mechanism of cytolysis by murine NK cells was analyzed using a variety of metabolic inhibitors that have proven informative in studying the lytic mechanism of CTL and the mechanism of histamine release by mast cells. Target cell binding occurred

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service