Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A4237

Sigma-Aldrich

Cefuroxime Axetil

Synonym(s):

[6R-[6α,7β(Z)]]-3-[[(Aminocarbonyl)oxy]methyl]-7-[[2-furanyl(methoxyimino)acetyl]amino]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid 1-(acetyloxy)ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H22N4O10S
CAS Number:
Molecular Weight:
510.47
MDL number:
UNSPSC Code:
51101500
PubChem Substance ID:
NACRES:
NA.85

assay

≥98% (HPLC)

form

powder

color

white to off-white

solubility

DMSO: 1 mg/mL

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

mode of action

cell wall synthesis | interferes

storage temp.

−20°C

SMILES string

O=C1[C@@H](NC(/C(C2=CC=CO2)=N\OC)=O)[C@]3([H])N1C(C(OC(C)OC(C)=O)=O)=C(COC(N)=O)CS3

InChI

1S/C20H22N4O10S/c1-9(25)33-10(2)34-19(28)15-11(7-32-20(21)29)8-35-18-14(17(27)24(15)18)22-16(26)13(23-30-3)12-5-4-6-31-12/h4-6,10,14,18H,7-8H2,1-3H3,(H2,21,29)(H,22,26)/b23-13-/t10?,14-,18-/m1/s1

InChI key

KEJCWVGMRLCZQQ-YJBYXUATSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Cefuroxime axetil is acetoxyethyl pro-drug cephalosporin with activity against some strains of β-lactamase producing bacteria. It is a broad spectrum antibiotic active against Gram-positive and Gram-negative bacteria.[1]

Other Notes

Keep container tightly closed in a dry and well-ventilated place.

pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ahmet Emre Eskazan et al.
Aesthetic plastic surgery, 36(2), 464-467 (2011-08-20)
Thrombotic thrombocytopenic purpura (TTP) or Moschcowitz's syndrome is characterized by platelet and von Willebrand factor (vWF) deposition in arterioles and capillaries throughout the body, which results in organ ischemia. The diagnostic pentad characterizing TTP consists of thrombocytopenia, microangiopathic hemolytic anemia
J B Bulitta et al.
Antimicrobial agents and chemotherapy, 53(8), 3462-3471 (2009-06-17)
Cefuroxime axetil is widely used to treat respiratory tract infections. We are not aware of a population pharmacokinetic (PK) model for cefuroxime axetil. Our objectives were to develop a semiphysiological population PK model and evaluate the pharmacodynamic profile for cefuroxime
Ramesh Bomma et al.
Pharmaceutical development and technology, 18(5), 1230-1237 (2012-02-22)
The objective of this investigation was to develop the cefuroxime axetil sustained-release floating tablets to prolong the gastric residence time and compare their pharmacokinetic behavior with marketed conventional tablets (Zocef). The floating tablets were developed using polymers like HPMC K4M
Dasa Cerar et al.
The American journal of medicine, 123(1), 79-86 (2010-01-28)
Controversy exists over the significance and even the existence of post-Lyme disease symptoms because of the high rate of similar background symptoms in the general population. A European, prospective clinical trial in which doxycycline and cefuroxime axetil were compared in
Inderbir Singh et al.
Acta poloniae pharmaceutica, 69(1), 87-93 (2012-05-12)
The direct compression suitability of active pharmaceutical ingredients could be studied by SeDeM diagram method. Cefuroxime axetil (CfA) and paracetamol (PCM) were employed for SeDeM studies as these powders are well-characterized and known to be particularly difficult with respect to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service