Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A4226

Sigma-Aldrich

Anti-ADAM-17, Propeptide Domain antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-A Disintegrin And Metalloproteinase-17, Anti-TACE, Anti-TNF-α Converting Enzyme

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

rat, pig, human

concentration

~1 mg/mL

technique(s)

western blot: 1:1,000

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... ADAM17(6868)
rat ... Adam17(57027)

General description

ADAM-17 or tumor necrosis factor converting enzyme (TACE) is known to regulate several cellular functions such as cell growth and movement. It has a varied array of substrates that include adhesion molecules, growth factors and their receptors, as well as many cytokines. Thus, ADAM-17 abnormalities have been associated with various disorders such as renal fibrosis, Alzheimer′s disease, rheumatoid arthritis, cancer, diabetes and cardiac disorders.
Rabbit anti-ADAM-17, propeptide domain antibody binds to human ADAM-17. The antibody reacts with human, porcine, and rat ADAM17 and does not react with other ADAMs. By immunoblotting against the reduced protein, the antibody recognizes a band of 110kDa from conditioned media or cell lysates.

Immunogen

synthetic peptide corresponding to the propeptide domain of human ADAM-17.

Application

Rabbit anti-ADAM-17, propeptide domain antibody can be used for western blotting (1:1,000) applications.

Physical form

Solution in 0.01 M phophate buffered saline containing 50% glycerol and 0.05% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Monika Gooz
Critical reviews in biochemistry and molecular biology, 45(2), 146-169 (2010-02-27)
This review focuses on the role of ADAM-17 in disease. Since its debut as the tumor necrosis factor converting enzyme (TACE), ADAM-17 has been reported to be an indispensible regulator of almost every cellular event from proliferation to migration. The

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service