A421080
DMT-dA(bz)-CPG Column
pore size 1000 Å, 0.2 μmol, configured for PerkinElmer
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352200
NACRES:
NA.51
type
for DNA synthesis
product line
Proligo Reagents
Proligo Reagents
assay
≥98.0% (Nucleoside Purity, reversed phase HPLC)
pore size
1000 Å
capacity
≥99 % (Coupling Efficiency)
λ
conforms (UV/VIS Identity)
compatibility
configured for PerkinElmer
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service