Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A3987

Sigma-Aldrich

Cefpodoxime proxetil

Synonym(s):

(RS)1(isopropoxycarbonyloxy) ethyl (+)-(6R,7R)-7-[2-(2-amino-4- thiazolyl)-2-{(Z)methoxyimino}acetamido]-3methoxymethyl-8-oxo-5-thia-1-azabicyclo [4.2.0]oct-2-ene- 2-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H27N5O9S2
CAS Number:
Molecular Weight:
557.60
UNSPSC Code:
51284139
NACRES:
NA.85

form

powder

storage temp.

−20°C

InChI

1S/C21H27N5O9S2/c1-9(2)33-21(30)35-10(3)34-19(29)15-11(6-31-4)7-36-18-14(17(28)26(15)18)24-16(27)13(25-32-5)12-8-37-20(22)23-12/h8-10,14,18H,6-7H2,1-5H3,(H2,22,23)(H,24,27)/b25-13-/t10?,14-,18-/m1/s1

InChI key

LTINZAODLRIQIX-FBXRGJNPSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Cefpodoxime proxetil is the esterified prodrug of cefpodoxime, a third-generation cephalosporin antibiotic with broad spectrum activity against most Gram-positive and Gram-negative bacteria. It inhibits bacterial cell wall synthesis by binding penicillin-binding proteins, preferentially penicillin binding protein 3, and resulting in the inhibition of peptidoglycan synthesis. Cefpodoxime is stable in the presence of most β-lactamase enzymes but inactivated by certain extended spectrum β-lactamases.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service