Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A3646

Sigma-Aldrich

Aminoacyl-tRNA Synthetase from Escherichia coli

buffered aqueous glycerol solution, ≥2,000 units/mg protein

Synonym(s):

ARS, tRNA-ligase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.26

form

buffered aqueous glycerol solution

Quality Level

specific activity

≥2,000 units/mg protein

shipped in

wet ice

storage temp.

−20°C

Application

Aminoacyl tRNA synthetase may be useful in the study of protein synthesis and regulation [1].

Biochem/physiol Actions

Aminoacyl tRNA synthetase attaches amino acids to their corresponding tRNA, which is essential for protein synthesis. Both classes of aminoacyl tRNA synthetases are multi-domain proteins [2].

Other Notes

Mixture of amino acid activating enzymes
EC Ligase sub-class 6.1.1

Quality

Crude

Unit Definition

One unit will activate and attach 1.0 picomole (10-12 mole) of labeled amino acid to tRNA in 10 min at pH 7.6 at 37 °C (amino acid used: L-arginine).

Physical form

Solution in 50% glycerol containing 10 mM Tris HCl, pH 7.2, 10 mM MgCl2, 30 mM 2-mercaptoethanol and 10 mM KCl

Analysis Note

Protein determined by biuret.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Skin Sens. 1

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C L Harris
Journal of bacteriology, 169(6), 2718-2723 (1987-06-01)
Aminoacyl-tRNA synthetases from several strains of Escherichia coli are shown to elute as a high-molecular-weight complex on 6% agarose columns (Bio-Gel A-5M). In contrast, very little synthetase activity was observed in such complexes on Sephadex G-200 columns, suggesting that these
Irem Avcilar-Kucukgoze et al.
Cell chemical biology, 27(7), 839-849 (2020-06-20)
Arginyltransferase ATE1 mediates posttranslational arginylation and plays key roles in multiple physiological processes. ATE1 utilizes arginyl (Arg)-tRNAArg as the donor of Arg, putting this reaction into a direct competition with the protein synthesis machinery. Here, we address the question of
Ziwei Liu et al.
PLoS genetics, 17(7), e1009675-e1009675 (2021-07-30)
Emerging evidence indicates that tRNA-derived small RNAs (tsRNAs) are involved in fine-tuning gene expression and become dysregulated in various cancers. We recently showed that the 22nt LeuCAG3´tsRNA from the 3´ end of tRNALeu is required for efficient translation of a
Lance W Rider et al.
The Journal of biological chemistry, 284(16), 10324-10333 (2009-01-14)
Dihydrouridine synthases (DUSs) are flavin-dependent enzymes that catalyze site-specific reduction of uracils in tRNAs. The mechanism of DUS 2 from Saccharomyces cerevisiae was studied. Previously published turnover rates for this DUS were very low. Our studies show that the catalytic
C R Woese et al.
Microbiology and molecular biology reviews : MMBR, 64(1), 202-236 (2000-03-08)
The aminoacyl-tRNA synthetases (AARSs) and their relationship to the genetic code are examined from the evolutionary perspective. Despite a loose correlation between codon assignments and AARS evolutionary relationships, the code is far too highly structured to have been ordered merely

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service