Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A3359

Sigma-Aldrich

Anti-Annexin I antibody,Mouse monoclonal

clone ANEX 5E4/1, purified from hybridoma cell culture

Synonym(s):

Anti-ANX1, Anti-ANXA1, Anti-Annexin A1, Anti-LPC1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

ANEX 5E4/1, monoclonal

form

buffered aqueous solution

mol wt

antigen ~37 kDa

species reactivity

human

packaging

antibody small pack of 25 μL

concentration

~1.5 mg/mL

technique(s)

immunocytochemistry: suitable
western blot: 4-8 μg/mL using A431 cells extract

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Annexin I belongs to the annexin superfamily. It is associated with cellular membranes in the presence of Ca2+. The protein has a large conserved core domain (~30kDa) required for calcium and phospholipid association. This gene is widely expressed in tissues.

Immunogen

mixture of native membrane proteins from human bone tissues, which includes whole native annexin proteins.

Application

Monoclonal Anti-Annexin I antibody produced in mouse has been used in western blotting.

Biochem/physiol Actions

Annexin I is an anti-inflammatory factor and plays an important role in neuroprotection. It is associated with the removal of apoptotic cells, protects from ischemia-like injury and prevents hepatic inflammation. Annexin I also protects against the breakdown of blood-brain-barrier in Alzheimer′s disease. It is involved in the homeostasis of innate immune system by mediating immune cells (for instance, neutrophils and macrophages).

Physical form

Solution in 0.01M phosphate buffered saline pH 7.4, containing 15 mM sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Downregulation of Annexin A1 is correlated with radioresistance in nasopharyngeal carcinoma.
Huang L
Oncology Letters, 12, 5229-5229 (2016)
W F Lo et al.
Journal of immunology (Baltimore, Md. : 1950), 162(9), 5398-5406 (1999-05-05)
Despite being a major group of intracellular pathogens, the role of class I-restricted T cells in the clearance of Gram-negative bacteria is not resolved. Using a murine typhoid model, a role for class I-restricted T cells in the immune response
A fragmented form of annexin A1 is secreted from C2C12 myotubes by electric pulse-induced contraction.
Goto-Inoue N
Molecular and Cellular Biochemistry, 411, 173-173 (2016)
Annexin A1 Is a Key Modulator of Mesenchymal Stromal Cell-Mediated Improvements in Islet Function.
Rackham CL
Diabetes, 65, 129-129 (2016)
Annexin A1 restores A?1-42 -induced blood-brain barrier disruption through the inhibition of RhoA-ROCK signaling pathway.
Park JC
Aging Cell, 16, 149-149 (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service