Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

A3254

Sigma-Aldrich

4-Amino-5-carboxy-2-ethylmercaptopyrimidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H9N3O2S
CAS Number:
Molecular Weight:
199.23
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

biological source

synthetic

form

liquid

storage temp.

−20°C

SMILES string

CCSc1ncc(C(O)=O)c(N)n1

InChI

1S/C7H9N3O2S/c1-2-13-7-9-3-4(6(11)12)5(8)10-7/h3H,2H2,1H3,(H,11,12)(H2,8,9,10)

InChI key

TYAKXNHFMATCHA-UHFFFAOYSA-N

Application

4-Amino-5-carboxy-2-ethylmercaptopyrimidine is a derivitized 2-mercaptopyrimidine.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W Ciesielski et al.
Talanta, 47(3), 745-752 (2008-10-31)
A new method for the determination of 2-mercaptopyrimidines, using their reaction with iodine in neutral and alkaline medium, is presented. The determinability range in the volumetric titration, in phosphate buffer with starch as an indicator, was found to be equal
Chun-Yu Lin et al.
Anticancer research, 22(2A), 937-942 (2002-05-17)
Free radicals and reactive oxygen metabolites have been implicated as important pathologic mediators in many clinical disorders and diseases. An efficient method of detecting the free radical scavenger effect is through xanthine oxidase (XO) inhibition. The inhibition efficiency on XO
Xiu-Hua Zhang et al.
Bioelectrochemistry (Amsterdam, Netherlands), 65(1), 41-46 (2004-11-04)
4-Amino-2-mercaptopyrimidine self-assembled monolayer (AMP SAMs/Au) was prepared on a gold electrode. The AMP SAMs/Au was characterized by using attenuated total reflection-fourier transform infrared (ATR-FTIR) and A.C. Impedance. The electrochemical behavior of brucine on AMP SAMs/Au was studied by cyclic voltammetry

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service