Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A2847

Sigma-Aldrich

Arg-Glu(EDANS)-[Asn670, Leu671]-Amyloid β/A4 Precursor Protein 770 Fragment 669-674-(DABCYL)Lys-Arg

≥96% (HPLC), solid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C91H129N25O25S
Molecular Weight:
2005.22
MDL number:
UNSPSC Code:
12352202

assay

≥96% (HPLC)

form

solid

storage temp.

−20°C

Gene Information

human ... APP(351)
mouse ... APP(11820)
rat ... APP(54226)

Amino Acid Sequence

Arg-Glu-EDANS-Glu-Val-Asn-Leu-Asp-Ala-Glu-Phe-Lys-DABCYL-Arg

General description

Fuorogenic substrate containing the β-secretase site of the APP Swedish mutation.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service