Key Documents
A2549
Sigma-Aldrich
Tape, autoclave indicator, Sigmaware
impregnated to show words Autoclaved
About This Item
UNSPSC Code:
55121611
NACRES:
NB.35
General description
  • Tape is impregnated to show words Autoclaved after 15 minutes of exposure at 250°F (121°C) in a steam autoclave
  • Tape is supplied in 500 in. (1,270 cm) roll on a 1 in. core (2.54 cm)
  • Tape Dimensions: W 3/4 in. (1.91 cm)
  • Product contains lead which under normal use, does not present a health hazard or safety risk
  • Product contains latex (dry natural rubber)
Legal Information
Sigmaware is a trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Customers Also Viewed
Slide 1 of 1