Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A2353

Sigma-Aldrich

ATF-2 human

≥90% (SDS-PAGE), recombinant, expressed in E. coli, N-terminal maltose binding protein tagged, lyophilized powder

Synonym(s):

ATF-2 (AA 20-109)-Maltose Binding Protein fusion Protein, Activating transcription factor-2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.56

biological source

human

Quality Level

recombinant

expressed in E. coli

assay

≥90% (SDS-PAGE)

form

lyophilized powder

mol wt

53 kDa

technique(s)

activity assay: suitable

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... ATF2(1386)

Physical form

Lyophilized powder in HEPES buffered salts containing NaCl, DTT, EGTA, Brij, trehalose, and protease inhibitors.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Herdegen et al.
Brain research. Brain research reviews, 28(3), 370-490 (1998-12-22)
This article reviews findings up to the end of 1997 about the inducible transcription factors (ITFs) c-Jun, JunB, JunD, c-Fos, FosB, Fra-1, Fra-2, Krox-20 (Egr-2) and Krox-24 (NGFI-A, Egr-1, Zif268); and the constitutive transcription factors (CTFs) CREB, CREM, ATF-2 and
C Livingstone et al.
The EMBO journal, 14(8), 1785-1797 (1995-04-18)
The ATF-2 transcription factor can mediate adenovirus E1A-inducible transcriptional activation. Deletion analysis has indicated that the N-terminal region of ATF-2 is essential for this response. Furthermore, the N-terminus can activate transcription in the absence of E1A when fused to a
Patrick J Metz et al.
Cell reports, 30(6), 1935-1950 (2020-02-13)
Alternative splicing is well understood to enhance proteome diversity as cells respond to stimuli. However, mechanistic understanding for how the spliceosome processes precursor messenger RNA (mRNA) transcripts to achieve template diversification is incomplete. We use recently developed enzymatic inhibitors of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service