Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A212130

Sigma-Aldrich

DMT-2′O-Methyl-rA(tac) Phosphoramidite

configured for ABI

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C53H64N7O9P
Molecular Weight:
974.09
UNSPSC Code:
12352200
NACRES:
NA.51

product line

Proligo Reagents

assay

≥98.0% (reversed phase HPLC)
≥99% (31P-NMR)

form

solid

technique(s)

oligo synthesis: suitable

λ

conforms (UV/VIS Identity)

compatibility

configured for ABI

nucleoside profile

base: adenosine
base protecting group: TAC
2' protecting group: methyl
5' protecting group: DMT
deprotection: fast

storage temp.

−20°C

SMILES string

COC1[C@H](OP(OCCC#N)N(C(C)C)C(C)C)[C@@H](COC(c2ccccc2)(c3ccc(OC)cc3)c4ccc(OC)cc4)O[C@H]1n5cnc6c(NC(=O)COc7ccc(cc7)C(C)(C)C)ncnc56

InChI

1S/C53H64N7O9P/c1-35(2)60(36(3)4)70(67-30-14-29-54)69-47-44(31-66-53(38-15-12-11-13-16-38,39-19-23-41(62-8)24-20-39)40-21-25-42(63-9)26-22-40)68-51(48(47)64-10)59-34-57-46-49(55-33-56-50(46)59)58-45(61)32-65-43-27-17-37(18-28-43)52(5,6)7/h11-13,15-28,33-36,44,47-48,51H,14,30-32H2,1-10H3,(H,55,56,58,61)/t44-,47-,48?,51-,70?/m1/s1

InChI key

JUYHIKKXVRPEAW-DLKIFXHQSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service