A21123-HH
DMT-2′Fluoro-dA(bz) Phosphoramidite
configured for ABI
Synonym(s):
DMT-2′Fluoro-dA(bz) Amidite
About This Item
biological source
non-animal source (no BSE/TSE risk)
product line
Proligo Reagents
assay
≥99% (31P-NMR)
≥99.0% (reversed phase HPLC)
form
powder
packaging
pkg of 1 × 0.25 g A211231-01
technique(s)
oligo synthesis: suitable
impurities
≤0.3% water content (Karl Fischer)
≤0.5% P(III) Impurities 100-169ppm (31P-NMR)
≤0.5% single Impurity (reversed phase HPLC)
≤3% residual Solvent content
color
white to off-white
λ
conforms (UV/VIS Identity)
suitability
conforms to structure for H-NMR
conforms to structure for LC-MS
compatibility
configured for ABI
nucleoside profile
base: deoxyadenosine
base protecting group: benzoyl
2' protecting group: fluoro
5' protecting group: DMT
deprotection: standard
storage temp.
−20°C
SMILES string
CC(C)N(C(C)C)P(OCCC#N)O[C@H]([C@H](O1)COC(C2=CC=C(OC)C=C2)(C3=CC=C(OC)C=C3)C4=CC=CC=C4)[C@@H](F)[C@@H]1N5C6=NC=NC(NC(C7=CC=CC=C7)=O)=C6N=C5
InChI
1S/C47H51FN7O7P/c1-31(2)55(32(3)4)63(60-27-13-26-49)62-42-39(61-46(40(42)48)54-30-52-41-43(50-29-51-44(41)54)53-45(56)33-14-9-7-10-15-33)28-59-47(34-16-11-8-12-17-34,35-18-22-37(57-5)23-19-35)36-20-24-38(58-6)25-21-36/h7-12,14-25,29-32,39-40,42,46H,13,27-28H2,1-6H3,(H,50,51,53,56)/t39-,40-,42-,46-,63?/m1/s1
InChI key
VCCMVPDSLHFCBB-MSIRFHFKSA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service