Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

A2005

Sigma-Aldrich

DL-Alaninol hydrochloride

≥98%

Synonym(s):

2-Amino-1-propanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H9NO · HCl
CAS Number:
Molecular Weight:
111.57
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.26

Product Name

DL-Alaninol hydrochloride, ≥98%, crystalline

assay

≥98%

form

crystalline

color

off-white to tan

SMILES string

Cl.CC(N)CO

InChI

1S/C3H9NO.ClH/c1-3(4)2-5;/h3,5H,2,4H2,1H3;1H

InChI key

RPDODBFXWRWFAH-UHFFFAOYSA-N

Biochem/physiol Actions

DL-Alaninol hydrochloride is a racemic mixture of D- and L-alaninol stereoisomers used as chiral modifiers on copper (Cu) surfaces.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wai Yeng Cheong et al.
Langmuir : the ACS journal of surfaces and colloids, 26(21), 16412-16423 (2010-10-27)
Temperature programmed desorption methods have been used to probe the enantioselectivity of achiral Cu(100), Cu(110), and Cu(111) single crystal surfaces modified by chiral organic molecules including amino acids, alcohols, alkoxides, and amino-alcohols. The following combinations of chiral probes and chiral
Stefano Turchini et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 10(11), 1839-1846 (2009-05-30)
A photoelectron circular dichroism (CD) study of the valence states of 2-amino-1-propanol (alaninol) in the gas phase is presented. The aim of the investigation is to reveal conformer population effects in the valence-state photoelectron spectrum. The experimental dispersion of the
G Contini et al.
Langmuir : the ACS journal of surfaces and colloids, 27(12), 7410-7418 (2011-05-25)
The formation of 2D chiral monolayers obtained by self-assembly of chiral molecules on surfaces has been widely reported in the literature. Control of chirality transfer from a single molecule to surface superstructures is a challenging and important aspect for tailoring
Paola Gori et al.
The journal of physical chemistry. B, 112(13), 3963-3970 (2008-03-11)
The adsorption of a single molecule of the D-enantiomer of alaninol (2-amino-1-propanol) on the surface of Cu(100) is investigated through density functional theory calculations. Different possible adsorption sites for D-alaninol are tested, and it is found that the most stable

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service