Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A1777

Sigma-Aldrich

Albumin, Naphthol Blue Black bovine

sedimentation marker

Synonym(s):

Amido Black 10B stained BSA, Black albumin

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.25

biological source

bovine

Quality Level

form

powder

UniProt accession no.

storage temp.

2-8°C

Gene Information

bovine ... ALB(280717)

Application

Used as a sedimentation marker for density gradient centrifugation.
Used as a sedimentation marker for density gradient centrifugation.[1]

Other Notes

BSA impregnated with naphthol blue-black dye.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amido black 10 B-stained bovine serum albumin (BSA) in density gradient centrifugation.
I Kouvonen et al.
Analytical biochemistry, 89(1), 306-308 (1978-08-15)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service