Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A1705

Sigma-Aldrich

S-(5′-Adenosyl)-L-homocysteine Hydrolase from rabbit erythrocytes

buffered aqueous glycerol solution, ≥10 units/mg protein (Lowry)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352200

form

buffered aqueous glycerol solution

specific activity

≥10 units/mg protein (Lowry)

foreign activity

Adenosine deaminase activity ≤1%

shipped in

dry ice

storage temp.

−70°C

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Enzyme in vertebrates which catabolizes S-adenosyl-L-homocysteine.

Unit Definition

One unit will hydrolyze 1.0 nanomole of S-adenosyl-L-homocysteine to adenosine and L-homocysteine per min at pH 7.2 at 37 °C.

Physical form

Solution in 25 mM Tris, pH 7.4, containing 1 mM dithiothreitol, 1 mM EDTA, and 20% glycerol.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ajit Shokar et al.
Bioorganic & medicinal chemistry letters, 22(12), 4203-4205 (2012-05-15)
In the present study, we carried out a structure-activity analysis in Trichomonas vaginalis of a series of adenosine and uridine analogues. The most potent compounds were found to be 2' and 3' modified adenosine analogues some of which are potent
John Walker et al.
Molecular and biochemical parasitology, 183(2), 166-176 (2012-03-28)
The rate of treatment failure to antileishmanial chemotherapy in Latin America is up to 64%. Parasite drug resistance contributes to an unknown proportion of treatment failures. Identification of clinically relevant molecular mechanisms responsible for parasite drug resistance is critical to
Tomáš Honzík et al.
Molecular genetics and metabolism, 107(3), 611-613 (2012-09-11)
We report on the seventh known patient with S-adenosylhomocysteine hydrolase (SAHH) deficiency presenting at birth with features resembling phosphomannomutase 2 (PMM2-CDG Ia) deficiency. Plasma methionine and total homocysteine levels were normal at 2 months and increased only after the 8th
Laura Giusti et al.
Molecular bioSystems, 8(4), 1088-1099 (2012-02-02)
Washing fluid (WF) from the colon rectal tract after surgical resection might represent a first step in obtaining a mixture of proteins derived from the secretion of tumoral epithelial cells potentially involved in the pathological progression of tissue. In this
M J Robins et al.
Journal of medicinal chemistry, 41(20), 3857-3864 (1998-09-28)
Treatment of a protected 9-(5, 6-dideoxy-beta-D-ribo-hex-5-ynofuranosyl)adenine derivative with silver nitrate and N-iodosuccinimide (NIS) and deprotection gave the 6'-iodo acetylenic nucleoside analogue 3c. Halogenation of 3-O-benzoyl-5,6-dideoxy-1, 2-O-isopropylidene-alpha-D-ribo-hex-5-enofuranose gave 6-halo acetylenic sugars that were converted to anomeric 1,2-di-O-acetyl derivatives and coupled with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service