Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A1601

Sigma-Aldrich

Anti-ASC/TMS1 antibody produced in rabbit

~0.5 mg/mL, affinity isolated antibody, PBS solution

Synonym(s):

Anti-Apoptosis-Associated Speck-like Protein containing a CARD/Target of Methylation-Induced Silencing

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

PBS solution

mol wt

antigen ~25 kDa

species reactivity

human

concentration

~0.5 mg/mL

technique(s)

western blot: 1 μg/mL

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... PYCARD(29108)

General description

ASC/TMS1 is a CARD domain containing protein that is expressed in a variety of human and mouse tissues. Ectopic expression of ASC/TMS1 is known to inhibit cell survival and cause cell death in human breast cancer tissues by activating caspase-9. Furthermore, overexpression of ASC/TMS1 can induce DNA fragmentation,,,. Rabbit anti-ASC/TMS1 recognizes human ASC/TMS1 (approximately 25 kDa) by immunoblotting.

Immunogen

synthetic peptide corresponding to amino acids 182-195 of human ASC.

Application

Rabbit anti-ASC/TMS1 antibody can be used for western blot assays at a concentration of 1 μg/ml.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phophate buffered saline containing 0.02% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Masumoto et al.
The Journal of biological chemistry, 274(48), 33835-33838 (1999-11-24)
The cytoskeletal and/or nuclear matrix molecules responsible for morphological changes associated with apoptosis were identified using monoclonal antibodies (mAbs). We developed mAbs against Triton X-100-insoluble components of HL-60 cells pretreated with all-trans retinoic acid. In particular, one mAb recognized a
J Masumoto et al.
Experimental cell research, 262(2), 128-133 (2001-01-05)
ASC (apoptosis-associated speck-like protein containing a CARD) was first identified as a cytosolic soluble protein that forms insoluble aggregates and enhances etoposide-induced apoptosis. We have cloned a murine ortholog of ASC (mASC) comprising 193 amino acids with a well-conserved pyrin
B B McConnell et al.
Cancer research, 60(22), 6243-6247 (2000-12-05)
Genetic and epigenetic alterations affecting proteins involved in apoptosis can contribute to the establishment and progression of cancer. Recently, our laboratory has isolated a novel gene, TMS1, that is aberrantly methylated and silenced in a significant proportion of human breast
K E Conway et al.
Cancer research, 60(22), 6236-6242 (2000-12-05)
Gene silencing associated with aberrant methylation of promoter region CpG islands is an acquired epigenetic alteration that serves as an alternative to genetic defects in the inactivation of tumor suppressor and other genes in human cancers. The hypothesis that aberrant

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service