Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A1477

Sigma-Aldrich

Angiostatin K1-3 human

≥90% (SDS-PAGE), recombinant, expressed in Pichia pastoris (without N-linked glycosylation)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200

recombinant

expressed in Pichia pastoris (without N-linked glycosylation)

assay

≥90% (SDS-PAGE)

form

liquid

impurities

endotoxin, tested

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PLG(5340)

Biochem/physiol Actions

A proteolytic fragment of plasminogen containing the first three kringle structures. A specific inhibitor of endothelial cell growth and angiogenesis. More active relative to kringles 1-4. Studies with recombinant angiostatin show that the tumor inhibitory activity resides in a fragment of K1-3.
A specific inhibitor of endothelial cell growth and angiogenesis. More active relative to kringles 1-4.

Physical form

Supplied in 0.15 M sodium chloride.

Legal Information

U.S. Patent Nos. 6,024,688; 5,861,372. Sold under license from CMCC.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service