Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A1476

Sigma-Aldrich

Anti-Acinus, C-Terminal antibody produced in rabbit

~0.5 mg/mL, affinity isolated antibody, PBS solution

Synonym(s):

Anti-Apoptotic Chromatin Condensation Inducer in the Nucleus

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

PBS solution

mol wt

antigen ~220 kDa

species reactivity

human

concentration

~0.5 mg/mL

technique(s)

western blot: 0.25-0.5 μg/mL

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ACIN1(22985)

General description

Acinus is a ubiquitous protein that induces apoptotic chromatin condensation in cells. This protein is cleaved by caspase-3 and an additional unknown protease that subsequently generates a small active peptide p17. This small peptide causes in vitro condensation of chromatin when it is added to purified nuclei. In humans and mice, there are three distinct splice variants, namely, AcinusL, AcinusS, and AcinusS′. Rabbit anti-acinus, C-terminal antibody binds to human acinus (approx. 220 kDa) by immunoblotting.

Immunogen

synthetic peptide corresponding to amino acids 1329-1341 of human AcinusL, 602-614 of human AcinusS′ and 571-583 of human AcinusS.

Application

Rabbit anti-acinus, C-terminal antibody can be used for western blot assays at a concentration of 0.25-0.5 μg/mL.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Condensed matter in cell death.
N Zamzami et al.
Nature, 401(6749), 127-128 (1999-09-18)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service