Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A112N8-HH

Sigma-Aldrich

NPPOC-dA(tac) RayDite

Synonym(s):

NPPOC-dA(tac) Amidite, NPPOC-dA(tac) Phosphoramidite

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C41H52N8O10P
CAS Number:
Molecular Weight:
847.87
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

type

for DNA synthesis

product line

Proligo Reagents

assay

≥96% (31P-NMR)
≥96.0% (reversed phase HPLC)

form

solid

packaging

pkg of 1 × 0.25 g A112N81-01
pkg of 1 × 1 g A112N01-01

λ

conforms (UV/VIS Identity)

suitability

conforms to structure for H-NMR
conforms to structure for NMR (13C-NMR)

shipped in

dry ice

storage temp.

−20°C

SMILES string

CC(C)N(C(C)C)P(OCCC#N)O[C@H]1C[C@@H](O[C@@H]1COC(=O)OCC(C)c2ccccc2[N+]([O-])=O)n3cnc4c(NC(=O)COc5ccc(cc5)C(C)(C)C)ncnc34

InChI

1S/C41H53N8O10P/c1-26(2)48(27(3)4)60(57-19-11-18-42)59-33-20-36(58-34(33)22-56-40(51)55-21-28(5)31-12-9-10-13-32(31)49(52)53)47-25-45-37-38(43-24-44-39(37)47)46-35(50)23-54-30-16-14-29(15-17-30)41(6,7)8/h9-10,12-17,24-28,33-34,36H,11,19-23H2,1-8H3,(H,43,44,46,50)/t28?,33-,34+,36+,60?/m0/s1

InChI key

VVRGITZKIKTVDE-DIHBZMSYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

RayDite is a trademark of Sigma-Aldrich Co. LLC

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service