Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A111P0-HH

Sigma-Aldrich

DMT-dA(bz) Pharmadite®

Synonym(s):

Adenosine, N-benzoyl-5′-O-[bis(4-methoxyphenyl)phenylmethyl]-2′-deoxy-, 3′-[2-cyanoethyl N,N-bis(1-methylethyl)phosphoramidite], Adenosine, N-benzoyl-5′-O-[bis(4-methoxyphenyl)phenylmethyl]-2′-deoxy-, 3′-[2-cyanoethyl bis(1-methylethyl)phosphoramidite], DMT-dA(bz) Phosphoramidite, N-benzoyl-5′-O-[bis(4-methoxyphenyl)phenylmethyl]-2′-deoxyadenosine, 3′-[2-cyanoethyl N,N-bis(1-methylethyl)phosphoramidite], N6-Benzoyl-5′-O-(4,4′-dimethoxytrityl)-2′-deoxyadenosine-3′-O-[O-(2-cyanoethyl)-N,N′-diisopropylphosphoramidite]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C47H52N7O7P
CAS Number:
Molecular Weight:
857.93
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
Pricing and availability is not currently available.

biological source

non-animal source (no BSE/TSE risk)

Quality Level

type

for DNA synthesis

product line

Proligo Reagents

assay

≥99.5% (reversed phase HPLC)
>99.5% (31P-NMR)

form

powder or granules

packaging

pkg of please inquire

technique(s)

oligo synthesis: suitable

impurities

>= 170ppm, none detected (S/N <2.5)
≤0.1% P(III) 100-169ppm (31P-NMR)
≤0.1% single unspecified Imprity (reversed phase HPLC)
≤0.3% dA2 (reversed phase HPLC, DMT-dA(bz) cyanoethyl-H-posphonate)
≤0.3% dA3 (reversed phase HPLC, DMT-dA(bz) phosphoramidate)
≤0.30 wt. % water content (Karl Fischer)
≤0.5% P(V) -25-99ppm (31P-NMR)
≤3.0 wt. % residual solvent content

color

white to off-white

solubility

soluble, clear

suitability

conforms to structure for H-NMR
conforms to structure for LC-MS

nucleoside profile

base: adenosine
base protecting group: benzoyl
5' protecting group: DMT
deprotection: standard

storage temp.

2-8°C

SMILES string

COc1ccc(cc1)C(OC[C@H]2O[C@H](C[C@@H]2OP(OCCC#N)N(C(C)C)C(C)C)n3cnc4c(NC(=O)c5ccccc5)ncnc34)(c6ccccc6)c7ccc(OC)cc7

InChI

1S/C47H52N7O7P/c1-32(2)54(33(3)4)62(59-27-13-26-48)61-40-28-42(53-31-51-43-44(49-30-50-45(43)53)52-46(55)34-14-9-7-10-15-34)60-41(40)29-58-47(35-16-11-8-12-17-35,36-18-22-38(56-5)23-19-36)37-20-24-39(57-6)25-21-37/h7-12,14-25,30-33,40-42H,13,27-29H2,1-6H3,(H,49,50,52,55)/t40-,41+,42+,62?/m0/s1

InChI key

GGDNKEQZFSTIMJ-AKWFTNRHSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Pharmadite is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service