Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A1097

Sigma-Aldrich

[Asn(4-aminobutyl)1,7,23,Gln(4-aminobutyl)3,11,22]-Amyloid β-Protein 1-40

≥90% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C218H355N65O52S
Molecular Weight:
4750.62
MDL number:
UNSPSC Code:
12352202

assay

≥90% (HPLC)

form

solid

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... APP(351)

Amino Acid Sequence

Asn-AB-Ala-Gln-AB-Phe-Arg-His-Asn-AB-Ser-Gly-Tyr-Gln-AB-Val-His-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Gln-AB-Asn-AB-Val-Gly-Ser-Asn-Lys-Gly-Ala-Ile-Ile-Gly-Leu-Met-Val-Gly-Gly-Val-Val

Biochem/physiol Actions

Modified amyloid beta-peptide that may be used to detect plaques in vitro or in vivo allowing for the early diagnosis of Alzheimer′s disease.
Modified amyloid beta-peptide to detect plaques in vitro or in vivo for early Alzheimer′s disease diagnosis.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service