Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A0601

Sigma-Aldrich

Activin Receptor-Like Kinase 1/Fc Chimera from mouse

>95% (SDS-PAGE), recombinant, expressed in NSO cells, lyophilized powder, suitable for cell culture

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.32

biological source

mouse

recombinant

expressed in NSO cells

assay

>95% (SDS-PAGE)

form

lyophilized powder

potency

20-80 ng/mL ED50

mol wt

50-55 kDa by SDS-PAGE (reducing)
calculated mol wt ~37.7 kDa

packaging

pkg of 100 μg

storage condition

avoid repeated freeze/thaw cycles (Do not store in a frost-free freezer.)

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

UniProt accession no.

storage temp.

−20°C

Gene Information

mouse ... Acvrl1(11482)

Biochem/physiol Actions

ALK-1 is a type I complex of the serine/threonine kinases. Type I and II receptors bind ligands in the TGF-β superfamily. Human and mouse ALK-1 share approximately 71% amino acid sequence identity in their extracellular domains.
Type I complex of the serine/threonine kinases; bind ligands in the TGF-β superfamily.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline.

Analysis Note

Can be used as a calibrator for ALK-1 antibodies.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Lux et al.
The Journal of biological chemistry, 274(15), 9984-9992 (1999-04-03)
Germ line mutations in one of two distinct genes, endoglin or ALK-1, cause hereditary hemorrhagic telangiectasia (HHT), an autosomal dominant disorder of localized angiodysplasia. Both genes encode endothelial cell receptors for the transforming growth factor beta (TGF-beta) ligand superfamily. Endoglin
P ten Dijke et al.
Oncogene, 8(10), 2879-2887 (1993-10-01)
Human cDNA clones encoding four novel putative transmembrane protein serine/threonine kinases, denoted activin receptor-like kinase (ALK) -1, -2, -3 and -4, were obtained using a polymerase chain reaction (PCR)-based strategy. The PCR primers were designed based upon the sequence similarity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service