Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A0438

Sigma-Aldrich

Alcohol Oxidase from Hansenula sp.

vacuum-dried powder, ≥0.6 units/mg solid

Synonym(s):

Alcohol:oxygen oxidoreductase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

fungus (Hansenula spp.)

Quality Level

form

vacuum-dried powder

specific activity

≥0.6 units/mg solid

mol wt

~600 kDa

storage temp.

−20°C

Application

Alcohol Oxidase from Hansenula sp. has been used:
  • in the immobilization on amperometric biosensor by glutaraldehyde cross linking[1]
  • as a component in pre-mixed hydrogels for the electrode preparation in the biosensors[2]
  • in the modification of screen printed Co-phthalocyanine/ Carbon electrode for alcohol biosensor preparation[3]

Alcohol oxidase is used to catalyze the oxidation of short-chain, primary, aliphatic alcohols to their respective aldehydes [4]. It may be used to study methanol metabolism is yeasts, such as Candida, Pichia, and Hansenula. It is useful to study protein translocation into peroxisomes.

Biochem/physiol Actions

Alcohol oxidase has the highest affinity for methanol. The affinity decreases with increasing chain length of the alkyl (R) group. Alcohol oxidase is primarily localized in the peroxisome but is also found in the cytoplasm [4].
Catalyzes the oxidization of primary alcohols to aldehydes.

Molecular Weight: ~600 kDa
Optimum pH: 8.5
pH Range: 5.5-8.5

Km values with the following substrates:
Allyl alcohol 33.3 mM
Benzyl alcohol 26.6 mM
Ethanol 16.2 mM
Formaldehyde 2.6 mM
Methanol 2.15 mM
N-Butanol 166 mM
n-Propanol 66 mM

Cofactor: FAD

Inhibitors: 1,4-butynediol (irreversible), propargyl alcohol (irreversible), cyclopropanol, cyclopropanone (suicide substrate), formaldehyde, H2O2, hydroxylamine, KBr, KCN, methanol (substrate inhibitor), NaN3, PCMB, propynal, urea, 4-chloromercuribenzoic acid

Unit Definition

One unit will oxidize 1.0 μmole of methanol to formaldehyde per min at pH 7.5 at 25 °C.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

An alcohol oxidase biosensor using PNR redox mediator at carbon film electrodes
Barsan MM and Brett CMA
Talanta, 74(5), 1505-1510 (2008)
Madalina M Barsan et al.
Talanta, 74(5), 1505-1510 (2008-03-29)
A new amperometric biosensor for ethanol monitoring has been developed and optimised. The biosensor uses poly(neutral red) (PNR), as redox mediator, which is electropolymerised on carbon film electrodes and alcohol oxidase (AlcOx) from Hansenula polymorpha as recognition element, immobilised by
B Vinet
Clinical chemistry, 33(12), 2204-2208 (1987-12-01)
This method for the specific determination of methanol in serum is based on the following two reactions: (formula; see text) Alcohol oxidase is not specific: it converts all lower alcohols to their corresponding aldehydes; however, formaldehyde dehydrogenase is specific and
Bienzyme biosensors for glucose, ethanol and putrescine built on oxidase and sweet potato peroxidase
Castillo J, et al.
Biosensors And Bioelectronics, 18(5-6), 705-714 (2003)
Hu Jin et al.
Applied biochemistry and biotechnology, 165(2), 559-571 (2011-05-14)
Improved porcine interferon-α (pIFN-α) production by recombinant Pichia pastoris was achieved by culture conditions optimization in a 5-l bioreactor. The results indicated that the pIFN-α concentration, specific methanol consumption rate, specific activities of alcohol oxidase, formaldehyde dehydrogenase, and formate dehydrogenase

Protocols

To measure alcohol oxidase activity, this assay uses 2,2′-azino-bis-(3-ethylbenzothiazoline-6-sulfonic acid) and a continuous spectrophotometric rate determination at 405 nm.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service