Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A0161

Sigma-Aldrich

Albumin, cysteinyl bovine

lyophilized powder

Synonym(s):

S-Cysteinyl-albumin

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352204

form

lyophilized powder

extent of labeling

<0.02 mol sulfhydryl per mol albumin

impurities

salt, essentially free

Storage temp.

2-8°C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The reaction of protein thiol groups with some disulphides.
T E ISLES et al.
The Biochemical journal, 88, 84-88 (1963-07-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service