Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

98379

Sigma-Aldrich

Nonidet P 40 Substitute solution

BioXtra, ampule, ~10% in H2O

Synonym(s):

Nonylphenyl-polyethylene glycol solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12161900
NACRES:
NA.25

description

non-ionic

product line

BioXtra

form

solution

quality

ampule

mol wt

average mol wt 680

concentration

~10% in H2O

aggregation number

~132

impurities

≤150 μM sulfhydryl oxidizing substances (carbonyls, peroxides)

refractive index

n20/D 1.348

CMC

0.059 mM (20-25°C)

transition temp

cloud point 45-50 °C

density

1.01 g/mL at 20 °C

cation traces

Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤50 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤500 mg/kg
Mg: ≤50 mg/kg
Mn: ≤5 mg/kg
Na: ≤500 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

application(s)

detection

InChI

1S/C19H32O3/c1-2-3-4-5-6-7-8-9-18-10-12-19(13-11-18)22-17-16-21-15-14-20/h10-13,20H,2-9,14-17H2,1H3

InChI key

BLXVTZPGEOGTGG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Nonidet is a trademark of Shell Chemical Co.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

L Dumont et al.
Molecular human reproduction, 23(11), 738-754 (2017-10-19)
Do freezing and in vitro culture procedures enhance the expression of proteins involved in apoptotic or autophagic pathways in murine pre-pubertal testicular tissue? IVM strongly modified apoptosis- and autophagy-related relative protein levels in mice testicular tissue whereas the impact of
Daniel J Smit et al.
Cells, 9(9) (2020-09-24)
Circulating tumor cells (CTCs) are cells shed from the primary tumor into the bloodstream. While many studies on solid tumor cells exist, data on CTCs are scarce. The mortality of cancer is mostly associated with metastasis and recent research identified

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service