storage temp.
2-8°C
General description
Features and Benefits
- Comprehensive, efficient screening
- Stringent purity standards
- Prefabricated reagents, application sheets, and observation sheets
- Low incidence of undesirable solubility effects
Components
Kit Components Only
- MES pH 6.5; 0.05M – Sodium chloride 0.1M – DTT 0.001M solution
- MES pH 6.5; 0.05M – Sodium chloride 0.2M – DTT 0.001M – PPS 1M solution
- HEPES Sodium salt pH 7.5; 0.05M – PPS 1M solution
- HEPES Sodium salt pH 7.5; 0.05M – Sodium chloride 0.1M solution
- TRIS hydrochloride pH 8.5; 0.05M – Sodium chloride 0.1M – PPS 1M solution
- TRIS hydrochloride pH 8.5; 0.05M – Sodium chloride 0.1M – DTT 0.001M solution
- TRIS hydrochloride pH 8.5; 0.05M – Sodium chloride 0.2M – DTT 0.001M – PPS 1M solution
- TRIS hydrochloride pH 8.5; 0.05M – L-Glutathione red. 0.005M – L-Glutathione ox. 0.001M – PPS 1M solution
- TRIS hydrochloride pH 8.5; 0.05M – Sodium chloride 0.2M – L-Glutathione red. 0.005M – L-Glutathione ox. 0.001M solution
- TRIS hydrochloride pH 8.5; 0.05M – Sodium chloride 0.2M – L-Glutathione red. 0.005M – L-Glutathione ox. 0.001M – PPS 1M solution
- MES pH 6.5; 0.05M – Sodium chloride 0.1M solution
- HEPES Sodium salt pH 7.5; 0.05M – 0.1M Sodium chloride, L-Glutathione red. 0.005M – L-Glutathione ox. 0.001M solution
- MES pH 6.5; 0.05M – Sodium chloride 0.2M – L-Glutathione red. 0.005M – L-Glutathione ox. 0.001M – PPS 1M solution
Kit Components Also Available Separately
- MES pH 6.5; 0.05M – Sodium chloride 0.1M – PPS 1M solution
- 82953MES pH 6.5; 0.05M – Sodium chloride 0.2M solutionSDS
- MES pH 6.5; 0.05M – Sodium chloride 0.2M – PPS 1M solution
- 73021MES pH 6.5; 0.05M – DTT 0.001M Solution solutionSDS
- 71023MES pH 6.5; 0.05M – DTT 0.001M – PPS 1M solutionSDS
- 83809MES pH 6.5; 0.05M – Sodium chloride 0.2M – DTT 0.001M solutionSDS
- MES pH 6.5; 0.05M – L-Glutathione red. 0.005M – L-Glutathione ox. 0.001M solution
- MES pH 6.5; 0.05M – L-Glutathione red. 0.005M – L-Glutathione ox. 0.001M , PPS 1M solution
- MES pH 6.5; 0.05M – Sodium chloride 0.1M – L-Glutathione red. 0.005M – L-Glutathione ox. 0.001M – PPS 1M solution
- MES pH 6.5; 0.05M – Sodium chloride 0.1M – L-Glutathione red. 0.005M – L-Glutathione ox. 0.001M solution
- MES pH 6.5; 0.05M – Sodium chloride 0.2M – L-Glutathione red. 0.005M – L-Glutathione ox. 0.001M solution
- 94717HEPES Sodium salt pH 7.5; 0.05M solutionSDS
- HEPES Sodium salt pH 7.5; 0.05M – Sodium chloride 0.1M – PPS 1M solution
- HEPES Sodium salt pH 7.5; 0.05M – Sodium chloride 0.2M solution
- HEPES Sodium salt pH 7.5; 0.05M – Sodium chloride 0.2M – PPS 1M solution
- HEPES Sodium salt pH 7.5; 0.05M – DTT 0.001M solution
- HEPES Sodium salt pH 7.5; 0.05M – DTT 0.001M – PPS 1M solution
- HEPES Sodium salt pH 7.5; 0.05M – Sodium chloride 0.1M – DTT 0.001M solution
- HEPES Sodium salt pH 7.5; 0.05M – Sodium chloride 0.1M – DTT 0.001M – PPS 1M solution
- HEPES Sodium salt pH 7.5; 0.05M – Sodium chloride 0.2M – DTT 0.001M solution
- HEPES Sodium salt pH 7.5; 0.05M – Sodium chloride 0.2M – DTT 0.001M – PPS 1M solution
- HEPES Sodium salt pH 7.5; 0.05M – L-Glutathione red. 0.005M – L-Glutathione ox. 0.001M solution
- HEPES Sodium salt pH 7.5; 0.05M – L-Glutathione red. 0.005M – L-Glutathione ox. 0.001M – PPS 1M solution
- HEPES Sodium salt pH 7.5; 0.05M – Sodium chloride 0.1M – L-Glutathione red. 0.005M – L-Glutathione ox. 0.001M – PPS 1M solution
- HEPES Sodium salt pH 7.5; 0.05M – Sodium chloride 0.2M – L-Glutathione red. 0.005M – L-Glutathione ox. 0.001M solution
- 80409HEPES Sodium salt pH 7.5; 0.05M – Sodium chloride 0.2M – L-Glutathione red. 0.005M – L-Glutathione ox. 0.001M – PPS 1M solutionSDS
- TRIS hydrochloride pH 8.5; 0.05M – Sodium chloride 0.1M solution
- TRIS hydrochloride pH 8.5; 0.05M – Sodium chloride 0.2M solution
- TRIS hydrochloride pH 8.5; 0.05M – Sodium chloride 0.2M – PPS 1M solution
- TRIS hydrochloride pH 8.5; 0.05M – DTT 0.001M solution
- 70603TRIS hydrochloride pH 8.5; 0.05M – DTT 0.001M – PPS 1M solutionSDS
- 81220TRIS hydrochloride pH 8.5; 0.05M – Sodium chloride 0.1M – DTT 0.001M – PPS 1M solutionSDS
- 72259TRIS hydrochloride pH 8.5; 0.05M – Sodium chloride 0.2M – DTT 0.001M solutionSDS
- TRIS hydrochloride pH 8.5; 0.05M – L-Glutathione red. 0.005M – L-Glutathione ox. 0.001M solution
- TRIS hydrochloride pH 8.5; 0.05M –Sodium chloride 0.1M – L-Glutathione red. 0.005M – L-Glutathione ox. 0.001M solution
- 72403TRIS hydrochloride pH 8.5; 0.05M – Sodium chloride 0.1M – L-Glutathione red. 0.005M – L-Glutathione ox. 0.001M – PPS 1M solutionSDS
Storage Class
10 - Combustible liquids
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service