Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

96468

Sigma-Aldrich

Zinc chloride

for molecular biology

Synonym(s):

Dichlorozinc

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ZnCl2
CAS Number:
Molecular Weight:
136.30
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

Molecular Biology
for molecular biology

vapor pressure

1 mmHg ( 428 °C)

assay

≥97.0% (AT)

reaction suitability

reagent type: catalyst
core: zinc

impurities

DNases, none detected
RNases, none detected
phosphatases, none detected
proteases, none detected
≤10 mg/kg total nitrogen (N)

pH

4-5.5 (25 °C, 1 M in H2O)

mp

293 °C (lit.)

solubility

H2O: 0.14 g/mL at 20 °C, clear, colorless

anion traces

sulfate (SO42-): ≤100 mg/kg

cation traces

As: ≤2 mg/kg
Ca: ≤100 mg/kg
Fe: ≤10 mg/kg
Pb: ≤50 mg/kg

suitability

in accordance for filter test

SMILES string

[Cl-].[Cl-].[Zn++]

InChI

1S/2ClH.Zn/h2*1H;/q;;+2/p-2

InChI key

JIAARYAFYJHUJI-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Other Notes

Deregulation of DNA polymerase β by sense and antisense RNA expression in mouse 3T3 cells[1].

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

target_organs

Respiratory system

Storage Class

8B - Non-combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Z Zmudzka et al.
Somatic cell and molecular genetics, 16(4), 311-320 (1990-07-01)
DNA polymerase beta (beta-pol) and its mRNA are maintained at constitutive levels during the cell cycle and during stages of cell growth in culture. To study biological consequences of variations in the level of this DNA repair enzyme and/or its
L Davies et al.
Cell death & disease, 2, e136-e136 (2011-04-01)
The activation and regulation of target genes by the tumour-suppressor p53 dictates the fate of a cell, with cell cycle arrest or apoptosis being two distinct outcomes. PERP (p53 apoptosis effector related to PMP-22), a p53 transcriptional target, is induced
Kaushik Nath et al.
Environmental science and pollution research international, 20(6), 4030-4045 (2012-12-06)
Utilization of agrowaste materials for the production of activated carbon, as an excellent adsorbent with large surface area, is well established industrially, for dephenolation of wastewater. In the present work, dried pods of Prosopis cineraria-a novel and low-cost agrowaste material-were
İbrahim Gülseren et al.
Food chemistry, 135(2), 770-774 (2012-08-08)
Whey protein isolate (WPI) nanoparticles were prepared using ethanol desolvation, and their capacity to incorporate ZnCl(2) was analysed. Desolvation was carried out at pH 9 and the volume of added ethanol was 0-3 times the volume of protein solution. The
Michele K Sankako et al.
Pharmacological reports : PR, 64(6), 1537-1546 (2013-02-15)
The aim of this study was to evaluate the changes in testicular function of rats due to cigarette smoke exposure and the possible mechanism by which zinc protects against these alterations. Male Wistar rats (60 days old) were randomly divided

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service