95260
Vitamin F from linseed oil
mixture of fatty acids
Synonym(s):
Essential fatty acids
About This Item
Recommended Products
biological source
synthetic
form
liquid
color
colorless to light yellow
refractive index
n20/D 1.473
storage temp.
2-8°C
General description
Biochem/physiol Actions
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service