Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

94504

Sigma-Aldrich

Dextran cross-linked G-50

20-80 μm

Synonym(s):

Sephadex® alternative

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.25

form

powder

particle size

20-80 μm

Application

Dextran cross-linked G-50, 20-80 μm may be used as an alternative to Sephadex® G-50 for gel filtration/size exclusion chromatography of biomolecules >30,000 m.w.

Other Notes

To gain a comprehensive understanding of our extensive range of Dextrans for your research, we encourage you to visit our Carbohydrates Category page.

Legal Information

Sephadex is a registered trademark of Cytiva

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

John S Philo
Current pharmaceutical biotechnology, 10(4), 359-372 (2009-06-13)
Although size exclusion chromatography (SEC) has been, and will continue to be, the primary analytical tool for characterization of the content and size distribution of non-particulate aggregates in protein pharmaceuticals, regulatory concerns are driving increased use of alternative and complementary
Gel-filtration chromatography.
Bollag DM.
Methods of Biochemical Analysis, 36, 1-9 (1994)
Gel filtration: size exclusion chromatography.
Lars Hagel
Methods of biochemical analysis, 54, 51-91 (2011-10-01)

Related Content

Dextrans are polysaccharides with molecular weights ≥1,000 Dalton, featuring a linear backbone of α-linked d-glucopyranosyl repeating units.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service