Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

93543

Sigma-Aldrich

N-Hydroxysuccinimide (NHS) activated MegaStokes dye 605

Synonym(s):

4-{2-[4-(Dimethylamino)phenyl]ethenyl]-1-{5-[(2,5-dioxo-1-pyrrolidinyl)oxy]-5-oxopentyl}pyridinium hexafluorophosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H28F6N3O4P
Molecular Weight:
567.46
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.32

form

lumps

fluorescence

λex 490 nm; λem 605 nm in ethanol

storage temp.

−20°C

SMILES string

F[P-](F)(F)(F)(F)F.CN(C)c1ccc(\C=C\c2cc[n+](CCCCC(=O)ON3C(=O)CCC3=O)cc2)cc1

InChI

1S/C24H28N3O4.F6P/c1-25(2)21-10-8-19(9-11-21)6-7-20-14-17-26(18-15-20)16-4-3-5-24(30)31-27-22(28)12-13-23(27)29;1-7(2,3,4,5)6/h6-11,14-15,17-18H,3-5,12-13,16H2,1-2H3;/q+1;-1

InChI key

UUHUBYOVKMOORG-UHFFFAOYSA-N

General description

The MegaStokes dyes for „click" biological labeling, licensed by LuminoChem are virtually covering the whole visible spectrum reaching the infra-red regime. Besides dyes that are being capable of participating in classical copper catalyzed 1,3-dipolar cycloaddition reaction with the counterparting function, we also provide dyes containing cyclooctyne moiety an alkyne derivative that enables copper free clicking to azides.
These dyes are noteworthy for their large Stokes-shift (120 or 125 nm). Dyes for this kind are exceptionally useful in fluorescence resonance electron transfer (FRET) applications as they do not interfere with the spectral bands of the second fluorophore a common problem in FRET technology. Azido sugars have been used for click-labeling of surface glycoproteins, recently. We have adapted this system to demonstrate the ability of our dyes to undergo bioorthogonal labeling reaction, efficiently. Prior to labeling, Chinese hamster ovary (CHO) cells were incubated with these MegaStockesdyes to test the possible cytostatic effects of these dyes. Experimental results have shown that neither dye is toxic up to 50 μM concentration.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Skin Irrit. 2

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

James A Richardson et al.
The Analyst, 138(13), 3626-3628 (2013-05-16)
The intense demand for fluorescence-based point of care (POC) DNA diagnostics is driving developments to reduce the size of instrumentation, imposing limitations on the optical hardware that can be included. Here we describe a combination of instrumentation and fluorogenic probes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service