Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

933B

Rabbit Negative Control Serum

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.24

form

aqueous solution

packaging

pkg of 15 mL ready to use (933B-02)
pkg of 200 mL ready to use (933B-05)

technique(s)

immunohistochemistry: suitable

shipped in

wet ice

storage temp.

2-8°C

General description

Negative Control Serum (Rabbit) is a solution of non-immune rabbit serum diluted in a buffered saline solution containing carrier protein.

Physical form

Solution containing rabbit negative serum, Tris buffer, pH 7.3-7.7, with 1% BSA and <0.1% sodium azide

Other Notes

For Technical Service please contact: 800-665-7284 or email: [email protected]

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service