Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

93309

Sigma-Aldrich

TRIS borate – EDTA buffer substance

for Northern and Southern blotting, powder

Synonym(s):

TBE buffer dry blend

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105319
NACRES:
NA.25

grade

Molecular Biology

product line

BioReagent

form

powder

quality

5× concentrate

impurities

DNases, none detected
RNases, none detected
phosphatases, none detected
proteases, none detected

color

white

pH

8.1-8.5

storage temp.

room temp

General description

TBE (Tris/Borate/EDTA) Buffer is commonly used in nucleic acid electrophoresis. This solution is effective under slightly basic conditions, which keeps DNA deprotonated, water-soluble and protected from degradation. This powder should be reconstituted in molecular biology grade water before use.

Application

TRIS borate – EDTA buffer substance is suitable for use as running buffer for agarose gel electrophoresis[1] and nucleic acid electrophoresis, which may be part of Northern and Southern blot hybridization procedures.

Components

The final 5x TBE liquid contains 0.445M TRIS borate (pH 8.3) and 0.01M EDTA in water.

pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Repr. 1B

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Detection of Chicken Meat Adulteration in Raw Hamburger Using
Polymerase Chain Reaction
M. Mehdizadeh
Journal of Food Quality (2014)

Protocols

Learn Northern and Southern blotting basics, with protocols and applications for macromolecule transfer to membrane supports.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service